(S)-2-Hydroxy-3-(4-hydroxyphenyl)propanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: (S)-3-(4-Hydroxyphenyl)-2-hydroxypropionic acid

A793870-100MG A793870-250MG
77105-534EA 416.94 USD
77105-534 77105-536
(S)-2-Hydroxy-3-(4-hydroxyphenyl)propanoic acid 98%
(S)-2-Hydroxy-3-(4-hydroxyphenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₄
MW: 182.17 g/mol
MDL Number: MFCD01632461
CAS Number: 23508-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR