3-Bromo-1H-indazole-6-carbonitrile 95%

Supplier: Ambeed
Warning

A788155-250MG A788155-1G
77105-512EA 59.42 USD
77105-512 77686-809
3-Bromo-1H-indazole-6-carbonitrile 95%
3-Bromo-1H-indazole-6-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₄BrN₃
MW: 222.04 g/mol
MDL Number: MFCD11007867
CAS Number: 1082041-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR