Methyl 6-chloro-5-(trifluoromethyl)nicotinate 95%

Supplier: Ambeed
Warning

A786053-100MG A786053-1G A786053-250MG A786053-5G
77105-498EA 31.83 USD
77105-498 77105-500 77105-502 77105-504
Methyl 6-chloro-5-(trifluoromethyl)nicotinate 95%
Methyl 6-chloro-5-(trifluoromethyl)nicotinate

Precaución: for Research Use Only

Formula: C₈H₅ClF₃NO₂
MW: 239.57 g/mol
MDL Number: MFCD22559931
CAS Number: 1360952-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR