4-Fluoro-2-(trifluoromethoxy)phenylacetic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoro-2-(trifluoromethoxy)phenylacetic acid, 2-(4-Fluoro-2-(trifluoromethoxy)phenyl)acetic acid

A786011-100MG A786011-1G A786011-250MG
77105-492EA 72.15 USD
77105-492 77105-494 77105-496
4-Fluoro-2-(trifluoromethoxy)phenylacetic acid 95%
4-Fluoro-2-(trifluoromethoxy)phenylacetic acid

Precaución: for Research Use Only

Formula: C₉H₆F₄O₃
MW: 238.14 g/mol
MDL Number: MFCD19687177
CAS Number: 1323966-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR