1,2-Bis(4-hydroxyphenyl)ethanone 98+%

Supplier: Ambeed
Warning

A785399-1G A785399-250MG
77105-488EA 65.78 USD
77105-488 77105-490
1,2-Bis(4-hydroxyphenyl)ethanone 98+%
1,2-Bis(4-hydroxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₃
MW: 228.24 g/mol
MDL Number: MFCD01102874
CAS Number: 3669-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR