tert-Butyl 3,5-difluorobenzoate 98%

Supplier: Ambeed
Warning

A785286-100G A785286-25G A785286-5G
77105-482EA 179.3 USD
77105-482 77105-484 77105-486
tert-Butyl 3,5-difluorobenzoate 98%
tert-Butyl 3,5-difluorobenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₂O₂
MW: 214.20 g/mol
MDL Number: MFCD11975648
CAS Number: 467442-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR