i-CRT3 95%

Supplier: Ambeed
Danger

Synonyms: 2-(((2-(4-ethylphenyl)-5-methyloxazol-4-yl)methyl)thio)-N-phenethylacetamide

A781835-10MG A781835-1MG A781835-25MG A781835-50MG A781835-5MG
77105-458EA 90.18 USD
77105-458 77105-460 77105-462 77105-464 77105-466
i-CRT3 95%
i-CRT3

Precaución: for Research Use Only

Formula: C₂₃H₂₆N₂O₂S
MW: 394.53 g/mol
MDL Number: MFCD14766288
CAS Number: 901751-47-1
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR