2-Bromo-4-methyl-5-nitroanisole 95+%

Supplier: Ambeed
Danger

A781511-1G A781511-250MG
77105-454EA 48 USD
77105-454 77105-456
2-Bromo-4-methyl-5-nitroanisole 95+%
2-Bromo-4-methyl-5-nitroanisole

Precaución: for Research Use Only

Formula: C₈H₈BrNO₃
MDL Number: MFCD27935373
CAS Number: 41447-19-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR