4-Fluorobenzaldehyde 95%

Supplier: Ambeed
Danger

A781101-25G A781101-100G
77105-452EA 5.31 USD
77105-452 77686-794
4-Fluorobenzaldehyde 95%
4-Fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₅FO
MW: 124.11 g/mol
MDL Number: MFCD00003378
CAS Number: 459-57-4
UN: 1989
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR