3-Bromo-5-methylthiophene-2-boronic acid 96%

Supplier: Ambeed
Warning

A780930-1G A780930-250MG A780930-5G
77105-446EA 166.57 USD
77105-446 77105-448 77105-450
3-Bromo-5-methylthiophene-2-boronic acid 96%
3-Bromo-5-methylthiophene-2-boronic acid

Precaución: for Research Use Only

Formula: C₅H₆BBrO₂S
MW: 220.88 g/mol
MDL Number: MFCD18311829
CAS Number: 1351859-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR