tetra-Chloroauric(III) acid trihydrate 99%
Supplier: Ambeed
|
Danger
|
Synonyms:
Gold(III) chloride trihydrate, Gold Chloride hydrate, Hydrogen tetrachloroaurate(III) trihydrate, Hydrogen tetrachloroaurate (III) trihydrate, tetra-Chloroauric acid trihydrate
A779595-1G
A779595-250MG
A779595-5G
A779595-10G
77105-438EA
84.88
USD
77105-438
77105-440
77105-442
77614-384
tetra-Chloroauric(III) acid trihydrate 99%
tetra-Chloroauric(III) acid trihydrate
Precaución: for Research Use Only
Formula:
HAuCl₄·3H₂O MW: 393.83 g/mol |
MDL Number:
MFCD00149904 CAS Number: 16961-25-4 UN: 3260 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...