2-Methoxy-6-quinolineboronic acid pinacol ester 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Methoxy-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
A779557-100MG
A779557-1G
A779557-250MG
77105-432EA
36.08
USD
77105-432
77105-434
77105-436
2-Methoxy-6-quinolineboronic acid pinacol ester 98%
2-Methoxy-6-quinolineboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₆H₂₀BNO₃ MW: 285.15 g/mol |
MDL Number:
MFCD18383956 CAS Number: 1201644-36-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...