4-(Trifluoromethoxy)benzoyl chloride 97%

Supplier: Ambeed
Danger

A778647-1G A778647-25G A778647-5G
77105-426EA 11 USD
77105-426 77105-428 77105-430
4-(Trifluoromethoxy)benzoyl chloride 97%
4-(Trifluoromethoxy)benzoyl chloride

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O₂
MDL Number: MFCD00052329
CAS Number: 36823-88-8
UN: 2920
ADR: 8,3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR