Z-D-Norleucine 99%

Supplier: Ambeed
Warning

Synonyms: (R)-2-(((Benzyloxy)carbonyl)amino)hexanoic acid, Z-D-Nle-OH, Cbz-D-Nle-OH

A778642-1G A778642-25G A778642-5G
77105-424EA 41 USD
77105-424 77901-539 77900-795
Z-D-Norleucine 99%
Z-D-Norleucine

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₄
MW: 265.309 g/mol
MDL Number: MFCD00063169
CAS Number: 15027-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR