trans,trans-1,5-Bis(4-fluorophenyl)-1,4-pentadien-3-one 98%

Supplier: Ambeed
Warning

A771517-1G A771517-5G
77105-412EA 19 USD
77105-412 77105-414
trans,trans-1,5-Bis(4-fluorophenyl)-1,4-pentadien-3-one 98%
trans,trans-1,5-Bis(4-fluorophenyl)-1,4-pentadien-3-one

Precaución: for Research Use Only

Formula: C₁₇H₁₂F₂O
MDL Number: MFCD00298485
CAS Number: 53369-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR