Methyl 4-formyl-2-methylbenzoate 95%

Supplier: Ambeed
Warning

A771050-1G A771050-5G
77105-402EA 129.43 USD
77105-402 77105-404
Methyl 4-formyl-2-methylbenzoate 95%
Methyl 4-formyl-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₃
MW: 178.18 g/mol
MDL Number: MFCD22380496
CAS Number: 74733-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR