Methyl 5-amino-4,6-dibromopicolinate 95%

Supplier: Ambeed
Danger

A770953-100MG A770953-1G A770953-250MG A770953-5G
77105-394EA 73.21 USD
77105-394 77105-396 77105-398 77105-400
Methyl 5-amino-4,6-dibromopicolinate 95%
Methyl 5-amino-4,6-dibromopicolinate

Precaución: for Research Use Only

Formula: C₇H₆Br₂N₂O₂
MW: 309.94 g/mol
MDL Number: MFCD28991855
CAS Number: 1935587-27-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR