Methyl 2-(3,5-difluorophenyl)acetate 95%

Supplier: Ambeed
Warning

A770530-10G A770530-1G A770530-5G
77105-388EA 378.75 USD
77105-388 77105-390 77105-392
Methyl 2-(3,5-difluorophenyl)acetate 95%
Methyl 2-(3,5-difluorophenyl)acetate

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
MW: 186.155 g/mol
MDL Number: MFCD12025024
CAS Number: 210530-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR