3-Cyano-4-methoxyphenylboronic acid 95%

Supplier: Ambeed
Warning

A770366-100MG A770366-1G
77105-384EA 112.46 USD
77105-384 77105-386
3-Cyano-4-methoxyphenylboronic acid 95%
3-Cyano-4-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BNO₃
MW: 176.965 g/mol
MDL Number: MFCD10697415
CAS Number: 911210-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR