1-Bromo-2-ethoxy-4-(trifluoromethyl)benzene 98+%

Supplier: Ambeed
Warning

A766881-100G A766881-1G A766881-25G A766881-5G
77105-362EA 285.39 USD
77105-362 77105-364 77105-366 77105-368
1-Bromo-2-ethoxy-4-(trifluoromethyl)benzene 98+%
1-Bromo-2-ethoxy-4-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₈BrF₃O
MW: 269.05 g/mol
MDL Number: MFCD20441886
CAS Number: 156605-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR