Potassium (3-carbamoylphenyl)trifluoroborate 96%

Supplier: Ambeed
Warning

A763301-1G A763301-250MG A763301-5G
77105-348EA 149 USD
77105-348 77105-350 77105-352
Potassium (3-carbamoylphenyl)trifluoroborate 96%
Potassium (3-carbamoylphenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₇H₆BF₃KNO
MW: 227.03 g/mol
MDL Number: MFCD09993284
CAS Number: 1323952-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR