2,3-Difluorophenol 98%

Supplier: Ambeed
Danger

A762398-10G A762398-1G A762398-5G A762398-25G
77105-340EA 11.67 USD
77105-340 77105-342 77105-344 77552-336
2,3-Difluorophenol 98%
2,3-Difluorophenol

Precaución: for Research Use Only

Formula: C₆H₄F₂O
MW: 130.09 g/mol
MDL Number: MFCD00010262
CAS Number: 6418-38-8
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR