4-Chlorophenyl trifluoromethanesulfonate 95%

Supplier: Ambeed
Warning

Synonyms: 4-Chlorophenyltriflate, 4-Chlorophenyl triflate

A760314-1G A760314-25G A760314-5G
77105-334EA 11 USD
77105-334 77105-336 77105-338
4-Chlorophenyl trifluoromethanesulfonate 95%
4-Chlorophenyl trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₇H₄ClF₃O₃S
MW: 260.618 g/mol
MDL Number: MFCD07784323
CAS Number: 29540-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR