4-Hydroxy-3-methyl-5h-furan-2-one 95%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxy-3-methylfuran-2(5H)-one

A760072-1G A760072-250MG A760072-5G
77105-328EA 81.69 USD
77105-328 77105-330 77105-332
4-Hydroxy-3-methyl-5h-furan-2-one 95%
4-Hydroxy-3-methyl-5h-furan-2-one

Precaución: for Research Use Only

Formula: C₅H₆O₃
MW: 114.099 g/mol
MDL Number: MFCD01860143
CAS Number: 516-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR