(2-Isopropylphenyl)hydrazine hydrochloride 95%

Supplier: Ambeed
Warning

A758918-1G A758918-5G
77105-324EA 81 USD
77105-324 77105-326
(2-Isopropylphenyl)hydrazine hydrochloride 95%
(2-Isopropylphenyl)hydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₅ClN₂
MDL Number: MFCD04966798
CAS Number: 58928-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR