BRD-7552 98%

Supplier: Ambeed
Warning

Synonyms: BRD7552, Ethyl 4-(((((2R,3R,4S,5R,6S)-4,5-bis((benzo[d][1,3]dioxol-5-ylcarbamoyl)oxy)-2-(hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3-yl)oxy)carbonyl)amino)benzoate

A757692-100MG A757692-50MG
77105-322EA 492 USD
77105-322 77900-545
BRD-7552 98%
BRD-7552

Precaución: for Research Use Only

Formula: C₃₃H₃₃N₃O₁₅
MDL Number: MFCD28133407
CAS Number: 1137359-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR