3-Fluoro-5-formylbenzeneboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde, Benzaldehyde, 3-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-, 3-Fluoro-5-formylphenylboronic acid pinacol ester

A757190-100MG A757190-1G A757190-250MG
77105-314EA 37.14 USD
77105-314 77105-316 77105-318
3-Fluoro-5-formylbenzeneboronic acid pinacol ester 97%
3-Fluoro-5-formylbenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BFO₃
MW: 250.07 g/mol
MDL Number: MFCD16996301
CAS Number: 1112209-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR