9-Benzylcarbazole-3-carboxaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 9-Benzyl-9H-carbazole-3-carbaldehyde, 9-Benzyl-3-formylcarbazole

A757024-100MG A757024-1G A757024-250MG
77105-306EA 24.41 USD
77105-306 77105-308 77105-310
9-Benzylcarbazole-3-carboxaldehyde 98%
9-Benzylcarbazole-3-carboxaldehyde

Precaución: for Research Use Only

Formula: C₂₀H₁₅NO
MW: 285.35 g/mol
MDL Number: MFCD00403522
CAS Number: 54117-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR