3-Ethylphenylacetylene 95%

Supplier: Ambeed
Warning

Synonyms: 1-Ethyl-3-Ethynylbenzene

A756890-250MG A756890-1G
77105-304EA 109.28 USD
77105-304 77614-342
3-Ethylphenylacetylene 95%
3-Ethylphenylacetylene

Precaución: for Research Use Only

Formula: C₁₀H₁₀
MW: 130.19 g/mol
MDL Number: MFCD11655757
CAS Number: 29074-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR