1,3-Bis[3,5-bis(trifluoromethyl)phenyl]thiourea 98%
Supplier: Ambeed
|
Warning
|
A756596-100MG
A756596-1G
A756596-250MG
A756596-5G
A756596-25G
77105-298EA
10
USD
77105-298
77105-300
77105-302
77614-334
77901-023
1,3-Bis[3,5-bis(trifluoromethyl)phenyl]thiourea 98%
1,3-Bis[3,5-bis(trifluoromethyl)phenyl]thiourea
Precaución: for Research Use Only
Formula:
C₁₇H₈F₁₂N₂S MW: 500.31 g/mol |
MDL Number:
MFCD00829878 CAS Number: 1060-92-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...