Di-1-naphthylmethanol 98%

Supplier: Ambeed
Warning

A756348-1G A756348-250MG A756348-5G
77105-286EA 19 USD
77105-286 77105-288 77105-290
Di-1-naphthylmethanol 98%
Di-1-naphthylmethanol

Precaución: for Research Use Only

Formula: C₂₁H₁₆O
MDL Number: MFCD11096937
CAS Number: 62784-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR