2-Amino-6-(Trifluoromethoxy)phenol 96%

Supplier: Ambeed
Warning

A751686-100MG A751686-1G A751686-250MG
77105-272EA 59.42 USD
77105-272 77105-274 77105-276
2-Amino-6-(Trifluoromethoxy)phenol 96%
2-Amino-6-(Trifluoromethoxy)phenol

Precaución: for Research Use Only

Formula: C₇H₆F₃NO₂
MW: 193.12 g/mol
MDL Number: MFCD18390129
CAS Number: 1261524-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR