N-(4-((4-Nitrobenzyl)oxy)phenyl)acetamide 95%

Supplier: Ambeed
Warning

A750713-100MG A750713-1G A750713-250MG A750713-5G
77105-258EA 5 USD
77105-258 77105-260 77105-262 77105-264
N-(4-((4-Nitrobenzyl)oxy)phenyl)acetamide 95%
N-(4-((4-Nitrobenzyl)oxy)phenyl)acetamide

Precaución: for Research Use Only

Formula: C₁₅H₁₄N₂O₄
MDL Number: MFCD00413778
CAS Number: 26258-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR