Methyl (E)-3-(2-bromophenyl)prop-2-enoate 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl (2E)-3-(2-bromophenyl)prop-2-enoate
A750356-1G
A750356-5G
77105-254EA
92.3
USD
77105-254
77105-256
Methyl (E)-3-(2-bromophenyl)prop-2-enoate 96%
Methyl (E)-3-(2-bromophenyl)prop-2-enoate
Precaución: for Research Use Only
Formula:
C₁₀H₉BrO₂ MW: 241.081 g/mol |
MDL Number:
MFCD11111009 CAS Number: 92991-89-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...