1,2-Diethoxy-3-fluorobenzene 95+%
Supplier: Ambeed
|
Warning
|
A750310-10G
A750310-1G
A750310-5G
77105-248EA
75
USD
77105-248
77105-250
77105-252
1,2-Diethoxy-3-fluorobenzene 95+%
1,2-Diethoxy-3-fluorobenzene
Precaución: for Research Use Only
Formula:
C₁₀H₁₃FO₂ |
MDL Number:
MFCD15146834 CAS Number: 226555-35-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...