4-Iodo-N-methylbenzamide 98%

Supplier: Ambeed
Warning

A749030-100MG A749030-1G A749030-250MG A749030-5G
77105-242EA 9 USD
77105-242 77105-244 77105-246 77686-757
4-Iodo-N-methylbenzamide 98%
4-Iodo-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₈INO
MDL Number: MFCD02945450
CAS Number: 89976-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR