trans-2-Bromocinnamic acid 98%

Supplier: Ambeed
Warning

A747884-100G A747884-1G A747884-25G A747884-500G A747884-5G
77105-226EA 119.89 USD
77105-226 77105-228 77105-230 77105-232 77105-234
trans-2-Bromocinnamic acid 98%
trans-2-Bromocinnamic acid

Precaución: for Research Use Only

Formula: C₉H₇BrO₂
MW: 227.06 g/mol
MDL Number: MFCD00016836
CAS Number: 7499-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR