2,3-Dimethylbenzene-1,4-diamine 95%

Supplier: Ambeed
Danger

A747030-1G A747030-250MG
77105-222EA 190.97 USD
77105-222 77105-224
2,3-Dimethylbenzene-1,4-diamine 95%
2,3-Dimethylbenzene-1,4-diamine

Precaución: for Research Use Only

Formula: C₈H₁₂N₂
MW: 136.19 g/mol
MDL Number: MFCD00058687
CAS Number: 5306-96-7
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR