3-Chloro-4-ethoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

A744936-1G A744936-25G A744936-5G
77105-206EA 15 USD
77105-206 77105-208 77105-210
3-Chloro-4-ethoxyphenylboronic acid 98%
3-Chloro-4-ethoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BClO₃
MDL Number: MFCD03701556
CAS Number: 279261-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR