2-Amino-3-(2-nitrophenyl)propanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: H-DL-Phe(2-NO₂)-OH

A744338-1G A744338-5G
77105-202EA 59.42 USD
77105-202 77105-204
2-Amino-3-(2-nitrophenyl)propanoic acid 98%
2-Amino-3-(2-nitrophenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O₄
MW: 210.18 g/mol
MDL Number: MFCD00508172
CAS Number: 35378-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR