1-(2,2-Difluoroethyl)piperazine hydrochloride 95%

Supplier: Ambeed
Warning

A743193-100MG A743193-1G A743193-250MG
77105-194EA 29.71 USD
77105-194 77105-196 77105-198
1-(2,2-Difluoroethyl)piperazine hydrochloride 95%
1-(2,2-Difluoroethyl)piperazine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃ClF₂N₂
MW: 186.63 g/mol
MDL Number: MFCD11499046
CAS Number: 184042-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR