2,4-Dibromo-5-fluoroaniline 98%

Supplier: Ambeed
Warning

A743165-100MG A743165-1G A743165-250MG A743165-5G
77105-186EA 20.16 USD
77105-186 77105-188 77105-190 77105-192
2,4-Dibromo-5-fluoroaniline 98%
2,4-Dibromo-5-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄Br₂FN
MW: 268.9091 g/mol
MDL Number: MFCD09878124
CAS Number: 1000578-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR