tert-Butyl 4-((((benzyloxy)carbonyl)amino)methyl)benzoate 95%

Supplier: Ambeed
Warning

A742531-1G A742531-100MG A742531-250MG A742531-5G
77105-184EA 92 USD
77105-184 77901-047 77901-878 77901-496
tert-Butyl 4-((((benzyloxy)carbonyl)amino)methyl)benzoate 95%
tert-Butyl 4-((((benzyloxy)carbonyl)amino)methyl)benzoate

Precaución: for Research Use Only

Formula: C₂₀H₂₃NO₄
MDL Number: MFCD16620768
CAS Number: 220851-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR