3-Iodo-N-methylbenzamide 97%

Supplier: Ambeed
Warning

A737425-1G A737425-250MG
77105-152EA 61.54 USD
77105-152 77105-154
3-Iodo-N-methylbenzamide 97%
3-Iodo-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₈INO
MW: 261.05 g/mol
MDL Number: MFCD02859818
CAS Number: 90434-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR