8-Nitro-7-quinolinecarboxaldehyde 98%

Supplier: Ambeed
Warning

A736136-100MG A736136-250MG
77105-148EA 24 USD
77105-148 77105-150
8-Nitro-7-quinolinecarboxaldehyde 98%
8-Nitro-7-quinolinecarboxaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₆N₂O₃
MDL Number: MFCD08436438
CAS Number: 101327-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR