2,6-Dichloro-4-fluoroaniline 98%

Supplier: Ambeed
Danger

A735512-100G A735512-1G A735512-25G A735512-5G A735512-10G
77105-134EA 158 USD
77105-134 77105-136 77105-138 77105-140 77614-262
2,6-Dichloro-4-fluoroaniline 98%
2,6-Dichloro-4-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄Cl₂FN
MDL Number: MFCD00142845
CAS Number: 344-19-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR