Cyclohexanecarboxaldehyde 95%

Supplier: Ambeed
Danger

A734117-25G A734117-5G
77105-120EA 15.92 USD
77105-120 77105-122
Cyclohexanecarboxaldehyde 95%
Cyclohexanecarboxaldehyde

Precaución: for Research Use Only

Formula: C₇H₁₂O
MW: 112.17 g/mol
MDL Number: MFCD00001457
CAS Number: 2043-61-0
UN: 1989
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR