3-Boc-amino-3-(4-bromophenyl)oxetane 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (3-(4-bromophenyl)oxetan-3-yl)carbamate

A733182-100MG A733182-1G A733182-250MG
77105-116EA 52 USD
77105-116 77105-118 77614-252
3-Boc-amino-3-(4-bromophenyl)oxetane 95%
3-Boc-amino-3-(4-bromophenyl)oxetane

Precaución: for Research Use Only

Formula: C₁₄H₁₈BrNO₃
MW: 328.21 g/mol
MDL Number: MFCD22628720
CAS Number: 1279090-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR