7-Methylpyrido[2,3-d]pyrimidine-2,4-diol 95%

Supplier: Ambeed
Warning

A733129-100MG A733129-1G A733129-250MG
77105-110EA 46 USD
77105-110 77105-112 77105-114
7-Methylpyrido[2,3-d]pyrimidine-2,4-diol 95%
7-Methylpyrido[2,3-d]pyrimidine-2,4-diol

Precaución: for Research Use Only

Formula: C₈H₇N₃O₂
MDL Number: MFCD23703400
CAS Number: 36075-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR