2,5-Dioxopyrrolidin-1-yl 3-(2-(3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanamido)ethoxy)propanoate 98%

Supplier: Ambeed
Warning

Synonyms: Mal-amido-PEG1-C2-NHS ester

A731713-250MG A731713-1G
77105-108EA 64.72 USD
77105-108 77686-732
2,5-Dioxopyrrolidin-1-yl 3-(2-(3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanamido)ethoxy)propanoate 98%
2,5-Dioxopyrrolidin-1-yl 3-(2-(3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanamido)ethoxy)propanoate

Precaución: for Research Use Only

Formula: C₁₆H₁₉N₃O₈
MW: 381.33 g/mol
MDL Number: MFCD20229604
CAS Number: 1260092-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR